Learn More
Thermo Scientific Dionex™ IonPac™ AS18-Fast columns 0.4x150mm
Catalog No. NC0314980 Shop All Thermo Scientific Products
Encompass
Use these shorter 150mm Thermo Scientific™ Dionex™ IonPac™ AS18-Fast columns for fast runs when monitoring inorganic anions in drinking water and wastewater samples. Dionex IonPac AS18 columns are high capacity hydroxide-selective anion-exchange columns.
Catalog No. NC0314980 Supplier Thermo Scientific Supplier No. 072062
Use these shorter 150mm Thermo Scientific™ Dionex™ IonPac™ AS18-Fast columns for fast runs when monitoring inorganic anions in drinking water and wastewater samples. Dionex IonPac AS18 columns are high capacity hydroxide-selective anion-exchange columns.