Learn More
Mac-Mod Avantor ACE UltraCore SuperC18, 95 , 2.5 m, 2.1 x 100 mm Solid-Core Column
SDP
Catalog No. 501051916
Encompass
Avantor ACE UltraCore™ SuperC18™ solid core (core shell) particle columns are ideal for demanding MS work offering improved MS signal intensity and maximum MS response. These columns combine high efficiency separations with low column back pressure. The Encapsulated Bonding Technology (EBT™) greatly increases the ligand coverage of the silica surface and effectively eliminates the effect of unbonded silanol groups on separations. This higher ligand coverage results in improved inertness, chromatographic performance and stability. Solid Core particle technology provides higher efficiencies per unit back pressure as compared to sub-2-micron UHPLC columns. Columns combine the proven advantages of UHPLC and HPLC columns with Solid Core Particle Technology. Solid-core columns can be used on both UHPLC and HPLC systems.
Catalog No. 50-105-1916 Supplier Mac-Mod Supplier No. CORE25A1002U
Avantor ACE UltraCore™ SuperC18™ solid core (core shell) particle columns are ideal for demanding MS work offering improved MS signal intensity and maximum MS response. These columns combine high efficiency separations with low column back pressure. The Encapsulated Bonding Technology (EBT™) greatly increases the ligand coverage of the silica surface and effectively eliminates the effect of unbonded silanol groups on separations. This higher ligand coverage results in improved inertness, chromatographic performance and stability. Solid Core particle technology provides higher efficiencies per unit back pressure as compared to sub-2-micron UHPLC columns. Columns combine the proven advantages of UHPLC and HPLC columns with Solid Core Particle Technology. Solid-core columns can be used on both UHPLC and HPLC systems.