Learn More
Medchemexpress LLC HY-124817 5mg Medchemexpress, Col003 CAS:328565-16-8 Purity:>98%
SDP
Catalog No. 501932597
Encompass_Preferred
Medchemexpress, HY-124817 5mg Col003 CAS:328565-16-8 Col003 is a potent inhibitor of Hsp47 , competitively binds to the collagen binding site on Hsp47 ( IC 50 , 1.8 μM), and inhibits collagen secretion by destabilizing the collagen triple helix [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2597 Supplier Medchemexpress LLC Supplier No. HY1248175MG
Medchemexpress, HY-124817 5mg Col003 CAS:328565-16-8 Col003 is a potent inhibitor of Hsp47 , competitively binds to the collagen binding site on Hsp47 ( IC 50 , 1.8 μM), and inhibits collagen secretion by destabilizing the collagen triple helix [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.