Learn More
Aviva Systems Biology Corp Col11a1 Antibody - middle region (ARP60014_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60014_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Col11a1. It was validated on Western Blot. . Alternate names: C530001D20Rik, choApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-918