Learn More
Aviva Systems Biology Corp COL11A2 Antibody - N-terminal region (ARP50265_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50265_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COL11A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human COL11A2. Alternate names: DFNA13, DFNB53, HKE5, PARP, STL3ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-60303