Learn More
Cell Signaling Technology COL1A1 Matched Antibody Pair, Human, Unconjugated, IgG, 1 Kit
SDP
Catalog No. 503133806
Encompass
The COL1A1 Matched Antibody Pair provides a optimized set of capture and detection antibodies designed for high-sensitivity immunoassay platforms. This pair is ideal for applications requiring custom antibody labeling, including conjugation with fluorophores, lanthanides, biotin, or beads, and is fully compatible with diverse platforms such as MSD, Quanterix Simoa, Luminex, and Alpha Technology. Each antibody binds to non-overlapping epitopes to ensure high performance in sandwich immunoassays.
  • Supplied as a complete kit containing two monoclonal antibodies, each at 100 μg.
  • Validated for human COL1A1 detection in ELISA applications.
  • Formulated in 1X PBS (pH 7.8), BSA-free, and azide-free.
  • Compatible with high-throughput platforms like AlphaScreen, AlphaLISA, LANCE, and HTRF.
  • Ready for custom conjugation to enable specialized assay development.
Catalog No. 50-313-3806 Supplier Cell Signaling Technology Supplier No. 86263S
The COL1A1 Matched Antibody Pair provides a optimized set of capture and detection antibodies designed for high-sensitivity immunoassay platforms. This pair is ideal for applications requiring custom antibody labeling, including conjugation with fluorophores, lanthanides, biotin, or beads, and is fully compatible with diverse platforms such as MSD, Quanterix Simoa, Luminex, and Alpha Technology. Each antibody binds to non-overlapping epitopes to ensure high performance in sandwich immunoassays.
  • Supplied as a complete kit containing two monoclonal antibodies, each at 100 μg.
  • Validated for human COL1A1 detection in ELISA applications.
  • Formulated in 1X PBS (pH 7.8), BSA-free, and azide-free.
  • Compatible with high-throughput platforms like AlphaScreen, AlphaLISA, LANCE, and HTRF.
  • Ready for custom conjugation to enable specialized assay development.