Learn More
Aviva Systems Biology Corp COL4A3 Antibody - middle region (ARP60006_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60006_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COL4A3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human COL4A3. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-915