Learn More
Medchemexpress LLC COL4A3 Human His 10ug
SDP
Catalog No. 5000183549
Encompass_Preferred
Recombinant human COL4A3 protein fragment (residues G1427-K1668) with an N-terminal 6xHis tag, produced in E. coli and supplied as a lyophilized powder. The material has an approximate molecular weight of 30.6 kDa, 91% purity by reducing SDS-PAGE, endotoxin <1 EU/μg, and is formulated in PBS with 6% trehalose. Store at -20°C; after reconstitution it is stable at 4°C for 1 week or at -20°C for longer when aliquoted with carrier protein.
  • Lyophilized format for long-term stability and convenient storage.
  • N-terminal 6xHis tag enables affinity purification and detection.
  • High purity suitable for biochemical and cell-based assays.
  • Low endotoxin level supports cell-based applications.
  • Formulated with trehalose to improve stability during lyophilization.
  • Recommended reconstitution at ≥100 μg/mL to maintain solubility.
Catalog No. 50-001-83549 Supplier Medchemexpress LLC Supplier No. HYP7214810UG
Recombinant human COL4A3 protein fragment (residues G1427-K1668) with an N-terminal 6xHis tag, produced in E. coli and supplied as a lyophilized powder. The material has an approximate molecular weight of 30.6 kDa, 91% purity by reducing SDS-PAGE, endotoxin <1 EU/μg, and is formulated in PBS with 6% trehalose. Store at -20°C; after reconstitution it is stable at 4°C for 1 week or at -20°C for longer when aliquoted with carrier protein.
  • Lyophilized format for long-term stability and convenient storage.
  • N-terminal 6xHis tag enables affinity purification and detection.
  • High purity suitable for biochemical and cell-based assays.
  • Low endotoxin level supports cell-based applications.
  • Formulated with trehalose to improve stability during lyophilization.
  • Recommended reconstitution at ≥100 μg/mL to maintain solubility.