Learn More
Medchemexpress LLC COL4A3 Human His 5ug
SDP
Catalog No. 5000183440
Encompass_Preferred
Recombinant human COL4A3 fragment expressed in E. coli and supplied as an N-terminal 6xHis-tagged lyophilized powder. The product corresponds to UniProt Q01955 residues G1427-K1668, has an approximate molecular weight of 30.6 kDa, and is provided with >90% purity by reducing SDS-PAGE. Reconstitute at ≥100 μg/mL in ddH2O; store lyophilized at -20°C, and aliquot and freeze for extended storage.
Catalog No. 50-001-83440 Supplier Medchemexpress LLC Supplier No. HYP721485UG
Recombinant human COL4A3 fragment expressed in E. coli and supplied as an N-terminal 6xHis-tagged lyophilized powder. The product corresponds to UniProt Q01955 residues G1427-K1668, has an approximate molecular weight of 30.6 kDa, and is provided with >90% purity by reducing SDS-PAGE. Reconstitute at ≥100 μg/mL in ddH2O; store lyophilized at -20°C, and aliquot and freeze for extended storage.