Learn More
Medchemexpress LLC Col4a3 protein, rat (HEK293, Fc) | 94.3% | 10 UG
SDP
Catalog No. 5000292491
Encompass_Preferred
Recombinant rat COL4A3 (collagen type IV alpha-3) Fc-fusion protein produced in HEK293 cells, supplied as a research reagent for biochemical and cell-based studies.
  • Recombinant rat COL4A3 Fc fusion protein.
  • Expressed in HEK293 mammalian cells to support native folding.
  • Human Fc tag for detection and purification applications.
  • High purity (94.3%).
  • Packaged as a 10 μg research reagent.
Catalog No. 50-002-92491 Supplier Medchemexpress LLC Supplier No. HYP7568310UG
Recombinant rat COL4A3 (collagen type IV alpha-3) Fc-fusion protein produced in HEK293 cells, supplied as a research reagent for biochemical and cell-based studies.
  • Recombinant rat COL4A3 Fc fusion protein.
  • Expressed in HEK293 mammalian cells to support native folding.
  • Human Fc tag for detection and purification applications.
  • High purity (94.3%).
  • Packaged as a 10 μg research reagent.