Learn More
Aviva Systems Biology Corp COL4A6 Antibody - middle region (ARP60012_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60012_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COL4A6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human COL4A6. Alternate names: MGC88184, DELXq22.3, CXDELq22.3ApplicationWB. Species reactivity: Human
Catalog No. 50-286-917