Learn More
Aviva Systems Biology Corp COL5A1 Antibody - N-terminal region (ARP60015_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60015_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COL5A1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human COL5A1. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-919