Learn More
Aviva Systems Biology Corp COL6A2 Antibody - C-terminal region (ARP45805_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45805_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against COL6A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human COL6A2. Alternate names: DKFZp586E1322, PP3610ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rat
Catalog No. 50-014-59140