Learn More
Aviva Systems Biology Corp COL8A2 Antibody - middle region (ARP60023_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60023_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COL8A2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human COL8A2. Alternate names: FECD, FECD1, FLJ00201, MGC116970, MGC116972, PPCD, PPCD2ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-921