Learn More
Sino Biological COL9A1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 12231T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human COL9A1 (rh COL9A1; Catalog#12231-H08H; P20849-3; Met1-Pro328). COL9A1 specific IgG was purified by Human COL9A1 affinity chromatography.
Catalog No. 50-224-8671