Learn More
Aviva Systems Biology Corp COL9A3 Antibody - C-terminal region (ARP60028_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60028_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COL9A3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human COL9A3. Alternate names: DJ885L7.4.1, EDM3, FLJ90759, IDD, MEDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-922