Learn More
Cayman Chemical Colagen Stndard 300uL
SDP
Catalog No. 502945843
Encompass_Preferred
A pyranocoumarin with diverse biological activities inhibits RANKL-induced osteoclastogenesis of primary mouse BMDMs at 10 UM and LPS-induced NO production in RAW 264.7 macrophages at 100 UM inhibits PAF-induced platelet aggregation in isolated rabbit platelet-rich plasma (IC50 50 UM) and contractions of isolated guinea pig ileum induced by histamine or leukotriene D4 (pD2s 4.84 and 4.34 respectively) decreases ovariectomy-induced bone loss in a mouse model of osteoporosis at 10 mg/kg reduces the number of tumors initiated by DMBA and promoted by TPA in a mouse two-stage model of skin carcinogenesis
Catalog No. 50-294-5843 Supplier Cayman Chemical Supplier No. 400557300 UL
A pyranocoumarin with diverse biological activities inhibits RANKL-induced osteoclastogenesis of primary mouse BMDMs at 10 UM and LPS-induced NO production in RAW 264.7 macrophages at 100 UM inhibits PAF-induced platelet aggregation in isolated rabbit platelet-rich plasma (IC50 50 UM) and contractions of isolated guinea pig ileum induced by histamine or leukotriene D4 (pD2s 4.84 and 4.34 respectively) decreases ovariectomy-induced bone loss in a mouse model of osteoporosis at 10 mg/kg reduces the number of tumors initiated by DMBA and promoted by TPA in a mouse two-stage model of skin carcinogenesis