Learn More
TARGETMOL CHEMICALS INC COLCEMID 10MG
SDP
Catalog No. 502577257
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Colcemid (NSC-3096) is an inhibitor of microtubule polymerization. It used to study embryonic cloning. purity: 99%
Catalog No. 50-257-7257 Supplier TARGETMOL CHEMICALS INC Supplier No. T1972010MG
Also available in 1 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Colcemid (NSC-3096) is an inhibitor of microtubule polymerization. It used to study embryonic cloning. purity: 99%