Learn More
AdipoGen Colcemid
SDP
Catalog No. 502562286
Encompass_Preferred
Chemical. CAS 477-30-5. Formula C21H25NO5. MW 371.4. Semisynthetic from colchicine Prod. No. AG-CN2-0048. Anti-cancer compound. Cell synchronization agent. Used as a drug in chemotherapy by synchronizing tumor cells at metaphase, the radiosensitive stage of the cell cycle. Microtubule assembly inhibitor. Depolymerizes microtubules and limits microtubule formation inactivates spindle fibre formation. Inhibits mitosis during cell division at metaphase by inhibiting spindle formation. Low concentrations affect microtubule dynamics and cell migration, while high concentrations promote microtubule detachment from microtubule organizing center. Apoptosis inducer. Thrombopoietic agent.
Catalog No. 50-256-2286 Supplier AdipoGen Supplier No. AGCR13567M005
Chemical. CAS 477-30-5. Formula C21H25NO5. MW 371.4. Semisynthetic from colchicine Prod. No. AG-CN2-0048. Anti-cancer compound. Cell synchronization agent. Used as a drug in chemotherapy by synchronizing tumor cells at metaphase, the radiosensitive stage of the cell cycle. Microtubule assembly inhibitor. Depolymerizes microtubules and limits microtubule formation inactivates spindle fibre formation. Inhibits mitosis during cell division at metaphase by inhibiting spindle formation. Low concentrations affect microtubule dynamics and cell migration, while high concentrations promote microtubule detachment from microtubule organizing center. Apoptosis inducer. Thrombopoietic agent.