Learn More
AdipoGen Colcemid (1 mg)
SDP
Supplier:  AdipoGen AGCR13567M001
Encompass_Preferred
Chemical. CAS: 477-30-5. Formula: C21H25NO5. MW: 371.4. Semisynthetic from colchicine (Prod. No. AG-CN2-0048). Anti-cancer compound. Cell synchronization agent. Used as a drug in chemotherapy by synchronizing tumor cells at metaphase, the radiosensitive stage of the cell cycle. Microtubule assembly inhibitor. Depolymerizes microtubules and limits microtubule formation (inactivates spindle fibre formation). Inhibits mitosis during cell division at metaphase by inhibiting spindle formation. Low concentrations affect microtubule dynamics and cell migration, while high concentrations promote microtubule detachment from microtubule organizing center. Apoptosis inducer. Thrombopoietic agent.
Catalog No. 50-205-3936