Learn More
Enzo Life Sciences Colcemid (1mg). CAS: 477-30-5
SDP
Supplier:  Enzo Life Sciences ALX430033M001
Encompass_Preferred
Depolymerizes microtubules and inhibits their formation. Induces apoptosis. Alternative name: Demecolcine. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol, chloroform or DMSO. Long Term Storage: +4°C.
Catalog No. 50-200-9160