Learn More
Selleck Chemical LLC COLCEMID 5MG
SDP
Supplier:  Selleck Chemical LLC E26315MG
Encompass_Preferred
Colcemid (Demecolcine), a derivative of colchicine, is a potent mitotic inhibitor which can incuces cell apoptosis and can be used for cancer research. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC2332973