Learn More
Cayman Chemical Colchicine, 64-86-8, 100 mg
SDP
Supplier:  Cayman Chemical 9000760100
Encompass_Preferred
An inhibitor of microtubule polymerization (IC50 = 3.2 µM); inhibits growth of MCF-7 human breast carcinoma cells (IC50 = 13 nM); inhibits neutrophil motility and activity when used at a dose of 5 µmol/kg in a mouse model of gout
Catalog No. NC0693537