Learn More
Enzo Life Sciences Colchicine (1g). CAS: 64-86-8
SDP
Catalog No. 502008917
Encompass_Preferred
Depolymerizes microtubules and inhibits tubulin polymerization. Induces apoptosis in a variety of cells. Purity: ≥94% (HPLC). Solubility: Soluble in DMSO (14mg/ml), 100% ethanol (7mg/ml), water (7mg/ml) or dimethylformamide (12mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-8917 Supplier Enzo Life Sciences Supplier No. ALX380033G001
Depolymerizes microtubules and inhibits tubulin polymerization. Induces apoptosis in a variety of cells. Purity: ≥94% (HPLC). Solubility: Soluble in DMSO (14mg/ml), 100% ethanol (7mg/ml), water (7mg/ml) or dimethylformamide (12mg/ml). Long Term Storage: +4°C.