Learn More
Selleck Chemical LLC Colchicine 25mg 64-86-8 Colchineos, Colchisol, Colcin
SDP
Supplier:  Selleck Chemical LLC S228425MG
Encompass_Preferred
Colchicine (Colchineos, Colchisol, Colcin) is a microtubule polymerization inhibitor with an IC50 of 3 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8081