Learn More
Selleck Chemical LLC Colchicine S2284-100mg
SDP
Catalog No. 503112852
Encompass_Preferred
Colchicine is a microtubule polymerization inhibitor with an IC50 of 3 nM
Catalog No. 50-311-2852 Supplier Selleck Chemical LLC Supplier No. S2284100MG
Colchicine is a microtubule polymerization inhibitor with an IC50 of 3 nM