Learn More
Medchemexpress LLC HY-16569 500mg , Colchicine CAS:64-86-8 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY16569500MG
Encompass_Preferred
Medchemexpress, HY-16569 500mg Colchicine CAS:64-86-8 Formula:C22H25NO6 Microtubule/Tubulin Purity:98% Colchicine is a tubulin inhibitor and a microtubule disrupting agent. Colchicine triggers apoptosis. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2412