Learn More
Medchemexpress LLC Colchicine | 64-86-8 | 99.9% | 399.44 | 100 MG
SDP
Catalog No. 5000391806
Encompass_Preferred
Colchicine is an orally active alkaloid that acts as a potent tubulin inhibitor and a microtubule disrupting agent. It is intended for research and analytical applications.
  • Inhibits microtubule polymerization with an IC50 of 3 nM.
  • Acts as a competitive antagonist of the α3 glycine receptors (GlyRs).
  • Prevents non-steroidal anti-inflammatory drug (NSAID)-induced small intestinal injury by inhibiting activation of the NLRP3 inflammasome.
  • Possesses extensive anti-inflammatory, immunosuppressive, and strong anti-fibrosis effects.
  • Has potential for gouty arthritis research.
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Can be used to induce Alzheimer's disease models.
Catalog No. 50-003-91806 Supplier Medchemexpress LLC Supplier No. HY16569100MG
Colchicine is an orally active alkaloid that acts as a potent tubulin inhibitor and a microtubule disrupting agent. It is intended for research and analytical applications.
  • Inhibits microtubule polymerization with an IC50 of 3 nM.
  • Acts as a competitive antagonist of the α3 glycine receptors (GlyRs).
  • Prevents non-steroidal anti-inflammatory drug (NSAID)-induced small intestinal injury by inhibiting activation of the NLRP3 inflammasome.
  • Possesses extensive anti-inflammatory, immunosuppressive, and strong anti-fibrosis effects.
  • Has potential for gouty arthritis research.
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Can be used to induce Alzheimer's disease models.