Learn More
Medchemexpress LLC Colchicine | 64-86-8 | 99.93% | 399.44 | 25 MG
SDP
Catalog No. 5000392801
Encompass_Preferred
Colchicine (Standard) is the analytical standard for Colchicine, an orally active alkaloid. It acts as a potent tubulin inhibitor and a microtubule disrupting agent, inhibiting microtubule polymerization with an IC50 of 3 nM. It also functions as a competitive antagonist of α3 glycine receptors (GlyRs) and can prevent NSAID-induced small intestinal injury. Colchicine demonstrates extensive anti-inflammatory, immunosuppressive, and anti-fibrosis effects, suggesting its potential for gouty arthritis research.
  • Analytical standard for research and analytical applications.
  • Potent tubulin inhibitor and microtubule disrupting agent.
  • Inhibits microtubule polymerization with an IC50 of 3 nM.
  • Competitive antagonist of the α3 glycine receptors (GlyRs).
  • Prevents NSAID-induced small intestinal injury.
  • Extensive anti-inflammatory, immunosuppressive, and anti-fibrosis effects.
  • Potential for gouty arthritis research.
Catalog No. 50-003-92801 Supplier Medchemexpress LLC Supplier No. 25MGHY16569R
Colchicine (Standard) is the analytical standard for Colchicine, an orally active alkaloid. It acts as a potent tubulin inhibitor and a microtubule disrupting agent, inhibiting microtubule polymerization with an IC50 of 3 nM. It also functions as a competitive antagonist of α3 glycine receptors (GlyRs) and can prevent NSAID-induced small intestinal injury. Colchicine demonstrates extensive anti-inflammatory, immunosuppressive, and anti-fibrosis effects, suggesting its potential for gouty arthritis research.
  • Analytical standard for research and analytical applications.
  • Potent tubulin inhibitor and microtubule disrupting agent.
  • Inhibits microtubule polymerization with an IC50 of 3 nM.
  • Competitive antagonist of the α3 glycine receptors (GlyRs).
  • Prevents NSAID-induced small intestinal injury.
  • Extensive anti-inflammatory, immunosuppressive, and anti-fibrosis effects.
  • Potential for gouty arthritis research.