Learn More
Medchemexpress LLC Colchicine-d3 | 1217625-62-1 | 99.2% | 402.46 | 5 MG
SDP
Catalog No. 5000391197
Encompass_Preferred
Colchicine-d3 is the deuterium labeled Colchicine. Colchicine is a tubulin inhibitor and a microtubule disrupting agent. It inhibits microtubule polymerization with an IC50 of 3 nM and is a competitive antagonist of the α3 glycine receptors (GlyRs). It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-91197 Supplier Medchemexpress LLC Supplier No. HY16569S15MG
Colchicine-d3 is the deuterium labeled Colchicine. Colchicine is a tubulin inhibitor and a microtubule disrupting agent. It inhibits microtubule polymerization with an IC50 of 3 nM and is a competitive antagonist of the α3 glycine receptors (GlyRs). It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS