Learn More
Crescent Chemical Co Inc COLCHICINE SOLUTION 10 5G/ML
SDP
Catalog No. 50249114
Encompass_Preferred
Colchicine solution 10 µg/ml sterile filtered In PBS. Size - 25ML Storage Conditions - -15 °C TO -25 °C Catalog Number - 47252.01
Catalog No. 50-249-114 Supplier Crescent Chemical Co Inc Supplier No. SE47252.01
Colchicine solution 10 µg/ml sterile filtered In PBS. Size - 25ML Storage Conditions - -15 °C TO -25 °C Catalog Number - 47252.01