Learn More
Cayman Chemical ColchIcosIde 10mg
SDP
Catalog No. 502952837
Encompass_Preferred
A glycoside form of 3-demethylcolchicine; does not displace [3H]-colchicine from rat brain tubulin in vitro at 25 μM; increases CYP2C9 and CYP2E1 levels in primary human hepatocytes at 1 and 10 μM
Catalog No. 50-295-2837 Supplier Cayman Chemical Supplier No. 1183110 MG
A glycoside form of 3-demethylcolchicine; does not displace [3H]-colchicine from rat brain tubulin in vitro at 25 μM; increases CYP2C9 and CYP2E1 levels in primary human hepatocytes at 1 and 10 μM