Learn More
Medchemexpress LLC Colchicoside | 477-29-2 | MFCD01736974 | 99.4% | 547.55 g/mol | C27H33NO11 | 5 MG
SDP
Catalog No. 5000262236
Encompass_Preferred
Colchicoside (CAS 477-29-2) is a tropolone alkaloid research compound reported to have anti-leishmanial activity. It is supplied as a solid reference material with defined molecular and purity characteristics for laboratory and analytical use.
  • High purity solid suitable for research and analytical applications.
  • Molecular formula C27H33NO11 and molecular weight 547.55 g/mol.
  • Available in small milligram pack sizes for assay and reference use.
  • Provided with product and safety documentation for laboratory handling.
  • Used as a biochemical standard and research reagent in parasitology studies.
Catalog No. 50-002-62236 Supplier Medchemexpress LLC Supplier No. HY1313005MG
Colchicoside (CAS 477-29-2) is a tropolone alkaloid research compound reported to have anti-leishmanial activity. It is supplied as a solid reference material with defined molecular and purity characteristics for laboratory and analytical use.
  • High purity solid suitable for research and analytical applications.
  • Molecular formula C27H33NO11 and molecular weight 547.55 g/mol.
  • Available in small milligram pack sizes for assay and reference use.
  • Provided with product and safety documentation for laboratory handling.
  • Used as a biochemical standard and research reagent in parasitology studies.