Learn More
Uline Cold Bricks 6/case
Catalog No. NC1239725
Encompass
Foam bricks maintain their shape either frozen or thawed. Freeze before use. Recommended for tight packaging situations. Nontoxic heavy-duty plastic pouch 5 mil poly. FDA compliant. Solid when frozen not flexible. May be reused if internal foam core is not broken or cracked. Can be used to prevent freezing if heated in a microwave. Heat on low setting for 30 second increments. Flip pack over every 30 seconds until warm. 100% Recyclable. Outer bag can be recycled. Inside floral foam will break down in a landfill.
Catalog No. NC1239725 Supplier Uline Supplier No. S14295
Foam bricks maintain their shape either frozen or thawed. Freeze before use. Recommended for tight packaging situations. Nontoxic heavy-duty plastic pouch 5 mil poly. FDA compliant. Solid when frozen not flexible. May be reused if internal foam core is not broken or cracked. Can be used to prevent freezing if heated in a microwave. Heat on low setting for 30 second increments. Flip pack over every 30 seconds until warm. 100% Recyclable. Outer bag can be recycled. Inside floral foam will break down in a landfill.