Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Major Pharmaceuticals Cold and Cough Tablets for High Blood Pressure

Catalog No. 22355132
Change view
Click to view available options
Quantity:
16 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-355-132 16 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-355-132 Supplier Major Pharmaceuticals Supplier No. MAJ 873602
Add to Cart
Add to Cart
This item is not returnable. View return policy

These Cough and Cold Tabs can be compared to Coricidin Cough & Cold Tabs. For high blood pressure and offer relief for coughs, runny nose and sneezing.

  • Compare to Coricidin
  • Red color
  • 16/Pack
  • Country of Origin: United States

Specifications

Product Type Cold and Cough Tablets
For Use With (Application) Relief for Coughs, Runny Nose and Sneezing
Color Red
Format Tablet
Quantity 16 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.