Learn More
Electron Microscopy Sciences Cold Embedding Molds, 1.5" diameter
SDP
Catalog No. 501926832
Encompass_Preferred
These 2 part molds may be used for all cold embedding epoxies. Comes with a detached cap for easy removal. The molds come in 3 different sizes to accommodate all of the samples you may have. The molds are made from HDPE and we recommend the use of mold release spray prior to use.
Diameter: 1½"
Catalog No. 50-192-6832 Supplier Electron Microscopy Sciences Supplier No. 5048121
These 2 part molds may be used for all cold embedding epoxies. Comes with a detached cap for easy removal. The molds come in 3 different sizes to accommodate all of the samples you may have. The molds are made from HDPE and we recommend the use of mold release spray prior to use.
Diameter: 1½"