Learn More
Thermo Scientific™ Cold Filterable Tryptone Soya Broth in BPCs
Description
Minimize prep time and maximize production for media fill trials with ready-to-use Thermo Scientific™ Cold Filterable Tryptone Soya Broth in BPCs. Simply connect the BioProcess Containers into your existing workflow to maximize process simulations and minimize secondary contamination.
Effective
- Achieve effective process simulations, with minimized risk of secondary contamination
- Just plug and play
- Minimize preparation time
- Satisfy regulatory compliance
- Save time, reduce labour costs
Easy-to-Use
- Innovative bag design and connector options for easy process integration
- Standard, ready-to-use, off-the-shelf product range
- No mixing, no sterilization, long shelf life at ambient storage
- No Mycoplasma contamination
- Qualified microbial recovery performance
- Comprehensive validation package for media, bag/components and filling process
- No filter blockages
- No mixing required
- No sterilization
- No WFI (water for injection) required
- No vessel cleaning
Pharmacopoeial-compliant Tryptone Soya Broth and Animal Derived Component Free (ADCF) vegetable peptone-based formulations are available.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
SmartSite™ is a trademark or registered trademark of the Cardinal Healthcare Inc.
ReadyMate™ is a trademark or registered trademark of the gE Healthcare BioSciences AB.
Specifications
Specifications
| Color | Clear, straw |
| Content And Storage | 2 to 25°C |
| Description | Cold Filterable Tryptone Soya Broth in BPCs |
| Format | Bioprocess Container |
| Product Type | Microbiology Media |
| Includes | Includes the following connectors: SmartSite, needle-free valve additional sampling port, Quick connect, +ö body (MPC), 1+ö triclamp and ReadyMate aseptic connector |
| Quantity | 20 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.