Learn More
Chemglass Life Sciences Condenser, Micro Cold Finger, 14/20 Inner Joint, 5mm Length Below Joint
SDP
Catalog No. CG1222A01
Encompass_Preferred
Condenser, Micro Cold Finger, 14/20 Inner Joint, 5mm Length Below Joint. Condenser for use in refluxing from 1 to 20mL of solvent in flasks having a capacity of 5 to 50mL. Minimal surface area provides for better yields while providing adequate cooling. The additional hose connection is provided to allow introduction of an inert gas such as nitrogen. Top hose connection is cooling inlet and middle hose connection is cooling outlet. Cold finger extends 5mm below the joint and is used on a flask with the listed joint size. Hose connection is 10mm at the largest serration. Design suggested by Dr. Kathleen Zandi, Pfizer Inc., Groton CT.
Catalog No. CG1222A01 Supplier Chemglass Life Sciences Supplier No. CG1222A01
Condenser, Micro Cold Finger, 14/20 Inner Joint, 5mm Length Below Joint. Condenser for use in refluxing from 1 to 20mL of solvent in flasks having a capacity of 5 to 50mL. Minimal surface area provides for better yields while providing adequate cooling. The additional hose connection is provided to allow introduction of an inert gas such as nitrogen. Top hose connection is cooling inlet and middle hose connection is cooling outlet. Cold finger extends 5mm below the joint and is used on a flask with the listed joint size. Hose connection is 10mm at the largest serration. Design suggested by Dr. Kathleen Zandi, Pfizer Inc., Groton CT.