Learn More
MSC Ansell AlphaTec Chemical-Resistant Coverall, Nomex, Size S, 5/Case
Catalog No. 5000686870
Encompass
The AlphaTec™ Breathable FR Coverall 66-677 is a reusable, chemical- and flame-resistant protective suit designed to provide a robust barrier against organic and inorganic chemicals, biological agents, and heat-related hazards. Constructed from a Nomex® trilaminate material, it offers protection against flash fires, arc flashes, and hot liquids while promoting breathability to reduce the risk of heat stress.
  • Provides chemical splash protection with penetration data for more than 70 chemicals.
  • Engineered with flame-resistant fabric to guard against burn injuries.
  • Features taped seams and double front storm flaps for leaktight protection.
  • Includes adjustable cuffs and hook-and-loop fastenings at the legs.
  • Certified to NFPA 1992, EN1149-5, ASTM F1671, ASTM F1891-02, and ASTM F1959-04 standards.
  • Rated at 9.0 ATPV and Hazard Risk Category (HRC) 2.
Catalog No. 50-006-86870 Supplier MSC Supplier No. 07917792
Add to Cart
Add to Cart
The AlphaTec™ Breathable FR Coverall 66-677 is a reusable, chemical- and flame-resistant protective suit designed to provide a robust barrier against organic and inorganic chemicals, biological agents, and heat-related hazards. Constructed from a Nomex® trilaminate material, it offers protection against flash fires, arc flashes, and hot liquids while promoting breathability to reduce the risk of heat stress.
  • Provides chemical splash protection with penetration data for more than 70 chemicals.
  • Engineered with flame-resistant fabric to guard against burn injuries.
  • Features taped seams and double front storm flaps for leaktight protection.
  • Includes adjustable cuffs and hook-and-loop fastenings at the legs.
  • Certified to NFPA 1992, EN1149-5, ASTM F1671, ASTM F1891-02, and ASTM F1959-04 standards.
  • Rated at 9.0 ATPV and Hazard Risk Category (HRC) 2.