Learn More
Microscope World SCHOTT KL 1600 LED Cold Light Source
SDP
Catalog No. NC2965460
Encompass_Preferred
This SCHOTT LED cold light source includes a two-position filter slider and provides continuous dimming from 0-100%.Color Temperature- 5600KLuminous Flux- Up to 6802 lm light flux.Voltage- 100~240V, 50-60Hz.Light Box- Intensity control 0-100%, and filter slot with 2-position filter slider.Light Box Dimensions / Weight- 114mm x 231mm x 137mm (4.5inch x 9.1inch x 5.4inch), 2.45kg (5.4 lb)Certifications- CEWarranty- 2 years
Catalog No. NC2965460 Supplier Microscope World Supplier No. KL1600
This SCHOTT LED cold light source includes a two-position filter slider and provides continuous dimming from 0-100%.Color Temperature- 5600KLuminous Flux- Up to 6802 lm light flux.Voltage- 100~240V, 50-60Hz.Light Box- Intensity control 0-100%, and filter slot with 2-position filter slider.Light Box Dimensions / Weight- 114mm x 231mm x 137mm (4.5inch x 9.1inch x 5.4inch), 2.45kg (5.4 lb)Certifications- CEWarranty- 2 years