Learn More
Perkin Elmer US LLC Cold Plasma Optimization Solution, 0.1 % HNO3, 250 mL
Catalog No. 502104219
Encompass_Preferred
Concentration of Element includes: Matrix Concentration Volume 0.1 % HNO3 10 µg/L: Li, Co, Rb, In, Cs, Pb 125 mL
Catalog No. 50-210-4219 Supplier Perkin Elmer US LLC Supplier No. N8151032
Concentration of Element includes: Matrix Concentration Volume 0.1 % HNO3 10 µg/L: Li, Co, Rb, In, Cs, Pb 125 mL