Learn More
Analytical Sales & Services Cold Sleeve Column Cooler/Heater
Catalog No. 50457253
Encompass
Cold Sleeve-30 Peltier Column¿ Operation Range: MµLtiple Pre-Set Temperature Settings through Graphical User Interface ¿ Accommodates a Wide Range of Columns up to 30cm in length and 7.8mm I.D. Compact Design¿ Fast Equilibration and Highly Responsive.Cold Sleeve¿ is a Peltier driven cooler/heater designed to hold a single column up to 30cm long and up to 7.8mm in diameter. The temperature range is 5¿ to 55¿C with 0.2¿C accuracy. A double insµLated cover maintains temperature stability within the chamber ¿ 0.2¿C. The high performance control unit incorporates state-of-the-art electronics with dual remote preset temperatures for advanced methods development work. RS232 control is Standard
Catalog No. 50-457-253 Supplier Analytical Sales & Services Supplier No. COLD SLEEVE30
Cold Sleeve-30 Peltier Column¿ Operation Range: MµLtiple Pre-Set Temperature Settings through Graphical User Interface ¿ Accommodates a Wide Range of Columns up to 30cm in length and 7.8mm I.D. Compact Design¿ Fast Equilibration and Highly Responsive.Cold Sleeve¿ is a Peltier driven cooler/heater designed to hold a single column up to 30cm long and up to 7.8mm in diameter. The temperature range is 5¿ to 55¿C with 0.2¿C accuracy. A double insµLated cover maintains temperature stability within the chamber ¿ 0.2¿C. The high performance control unit incorporates state-of-the-art electronics with dual remote preset temperatures for advanced methods development work. RS232 control is Standard