Learn More
Andwin Scientific COLD STORAGE ADHESIVE SEALING
SDP
Catalog No. NC0249356
Encompass_Preferred
With a biologically inert adhesive, the cold storage adhesive sealing foil features a secure seal from -80 to 130C (-112 to 266F). This product is also able to lessen the chance of evaporation and is easily pierceable using a manual or automated pipettor tips or robotic probes. With these features, the product eliminates the probability of the generation of aerosols or cross-contamination.
Catalog No. NC0249356 Supplier Andwin Scientific Supplier No. 89049034
With a biologically inert adhesive, the cold storage adhesive sealing foil features a secure seal from -80 to 130C (-112 to 266F). This product is also able to lessen the chance of evaporation and is easily pierceable using a manual or automated pipettor tips or robotic probes. With these features, the product eliminates the probability of the generation of aerosols or cross-contamination.