Learn More
Unity Lab Services™ Cold Storage Equipment Requalification Service with Temperature Mapping
Shop All Thermo Scientific ProductsDescription
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications.
- Verifies important equipment functions and helps ensure that the equipment operates as expected by the manufacturer and conforms to standards and requirements.
- Key tests include door ajar, power failure, and temperature alerts.
- Temperature mapping (TM): A mapping study that establishes the temperature distribution within the chamber being mapped and locates hot and cold spots, which provides a view of the equipments performance.
- Temperature measurements will be performed in the air or with simulated products loaded in a chamber, with a data collection interval configurable to customer needs. The collected data provide essential information, helping ensure all material is correctly stored within its labeled temperature range.
- TM includes one open door temperature recovery test, one power failure test, minimum and maximum values along with averages, a temperature graph over time, and detailed reports that include raw probe data.
- Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
Specifications
Specifications
| For Use With (Equipment) | Cold Storage Equipment |
| Product Type | Requalification Service |