Learn More
Unity Lab Services™ Cold Storage Equipment Requalification Service
Catalog No. 1673545 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Cold Storage Equipment
Catalog No. For Use With (Equipment)
16-735-45 Cold Storage Equipment
1 options
Catalog No. 16-735-45 Supplier Unity Lab Services™ Supplier No. RQCOLD
Add to Cart
Add to Cart
Requalification Services for Cold Storage Equipment (documentation included)
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications.
  • Verifies important equipment functions and helps ensure that the equipment operates as expected by the manufacturer and conforms to standards and requirements.
  • Key tests include door ajar, power failure, and temperature alerts.
  • Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cold Storage Equipment
Product Type Requalification Service