Learn More
USA Scientific Inc Cold Storage foil, sterile
Catalog No. NC9553205
Encompass
Aluminum sealing foil for long term cold storage at -80C, sterile, 50/pack
Catalog No. NC9553205 Supplier USA Scientific Inc Supplier No. 29320110
Aluminum sealing foil for long term cold storage at -80C, sterile, 50/pack