Learn More
BUCHI Cold Trap Condenser Glass Assembly
SDP
Catalog No. 05405030 Shop All BUCHI Products
Change view
Click to view available options
Volume (Metric):
1000 mL
Catalog No. Volume (Metric)
05-405-030 1000 mL
1 options
Catalog No. 05-405-030 Supplier BUCHI Supplier No. 040640
Add to Cart
Add to Cart
Cold Trap Condenser Glass Assembly for Rotavapor R-300, R-100, R-2XX, R II, R-3, 500 cm2. For distillations of low-boiling point solvents, dry ice is needed. Includes a 1 liter receiving flask, the required tubings, and a ball joint clamp.
  • Includes 1 liter receiving flask, the required tubings, and a ball joint clamp
  • Does NOT include evaporating flask, vacuum gasket, vapor duct, and condenser holder
  • For use with Rotavapor R-300, R-100, R-2XX, R II, R-3
Specifications
Material Borosilicate Glass (3.3)
Type Condensor
For Use With (Application) Dry Ice, Condenser Assembly
For Use With (Equipment) Rotavpor™ R-100, R-3, R II, R-300, R-210, R-215
Volume (Metric) 1000 mL