Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ready America Cold Weather Survival Kit
SDP

Catalog No. 17985127
Change view
Click to view available options
Weight (English):
11 lb.
15 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (English)
17-985-127 11 lb.
17-985-128 15 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-985-127 Supplier Ready America Supplier No. 70400
Only null left
Add to Cart
Add to Cart

A 3 day Backpack containing essential items for winter storms.

  • Be prepared for what Mother Nature dishes out with a Cold Weather Emergency Kit by Ready America
  • Our kit contains essential items such as lifesaving food and water, survival blankets and body warmers, tools to dig your vehicle out of the snow and more
  • Available for one and two person kit for 3 day backpack

Specifications

Product Type Survival Kit
Dimensions (L x W x H) 12 x 9 x 16 in.
Includes Survival Blanket
Weight (English) 11 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.