Learn More
Cable Ties And More Specialized Nylon Cable Ties Suited For -40º F Temperatures, 100 Pcs Per Pack, 11"
SDP
Supplier:  Cable Ties And More CTCW11
Encompass_Preferred
This item has a minimum order quantity of 18 per supplier requirements
Specialized Nylon Cable Ties Suited For -40º F Cold Temperatures.100 Pcs Per Pack. 11" long.50 Lbs Tensile Strength.Tested For Application Performance at -20°C/-4°F Material Rated At -40°C/-40°F Per ISO180
Catalog No. NC1917302