Learn More
Cable Ties And More COLD WEATHER ZIP TIES 14"CS100
SDP
Catalog No. NC1917303
Encompass_Preferred
Specialized Nylon Cable Ties Suited For -40º F Cold Temperatures.100 Pcs Per Pack. 14" long.50 Lbs Tensile Strength.Tested For Application Performance at -20°C/-4°F Material Rated At -40°C/-40°F Per ISO180
Catalog No. NC1917303 Supplier Cable Ties And More Supplier No. CTCW14
Specialized Nylon Cable Ties Suited For -40º F Cold Temperatures.100 Pcs Per Pack. 14" long.50 Lbs Tensile Strength.Tested For Application Performance at -20°C/-4°F Material Rated At -40°C/-40°F Per ISO180